Product Details
- SNP ID
-
rs12737547
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:156646650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGACCTAGAGGTAGGGCTGCAGG[A/G]CCCTGGCCCCTGGCCCCTGGCCCCT
- Phenotype
-
MIM: 600347
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BCAN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113490499] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCAN
- Gene Name
- brevican
View Full Product Details