Product Details

SNP ID
rs11121562
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:10401446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAGTGCAGTGATAGCTCAGGG[A/G]AGGCTGAGCATTCCTTTCTAGAGGA
Phenotype
MIM: 172200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGD PubMed Links

Gene Details

Gene
PGD
Gene Name
phosphogluconate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304451.1 Intron NP_001291380.1
NM_001304452.1 Intron NP_001291381.1
NM_002631.3 Intron NP_002622.2

View Full Product Details