Product Details

SNP ID
rs11080328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35248204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAAGGATGATCTGGGCCTTTAC[G/T]GGCTATTTTGGGGTACATTTGACTT
Phenotype
MIM: 614952
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371932 PubMed Links

Gene Details

Gene
LOC105371932
Gene Name
uncharacterized LOC105371932
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017025492.1 Intron XP_016880981.1
Gene
SLFN5
Gene Name
schlafen family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144975.3 Intron NP_659412.3
XM_005257934.3 Intron XP_005257991.1

View Full Product Details