Product Details

SNP ID
rs11652356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40391147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGCTCACTGCAGCCTTGAACTC[C/T]TGGCCTCAAGCAATCCTCTTGTTTT
Phenotype
MIM: 126430
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOP2A PubMed Links

Gene Details

Gene
TOP2A
Gene Name
topoisomerase (DNA) II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001067.3 Intron NP_001058.2
XM_005257632.1 Intron XP_005257689.1
XM_011525165.1 Intron XP_011523467.1

View Full Product Details