Product Details

SNP ID
rs6943090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73332645 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCTGGGAATGTTCTGTGTTTTGA[C/T]GTGGTTCTTGGGTACATGGGTGTGT
Phenotype
MIM: 604839 MIM: 612548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP6 PubMed Links

Gene Details

Gene
FKBP6
Gene Name
FK506 binding protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135211.2 Intron NP_001128683.1
NM_001281304.1 Intron NP_001268233.1
NM_003602.4 Intron NP_003593.3
XM_006716153.2 Intron XP_006716216.1
XM_017012741.1 Intron XP_016868230.1
XM_017012742.1 Intron XP_016868231.1
Gene
TRIM50
Gene Name
tripartite motif containing 50
There are no transcripts associated with this gene.

View Full Product Details