Product Details

SNP ID
rs11465316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75811711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGAAACAGGAAAATTAGCTCTTC[A/C]CCCCATCACTGTGGCTTCTCCAGGC
Phenotype
MIM: 602495
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCL24 PubMed Links

Gene Details

Gene
CCL24
Gene Name
C-C motif chemokine ligand 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002991.2 712 Intron NP_002982.2
XM_011516459.2 712 UTR 3 XP_011514761.1
XM_011516460.2 712 UTR 3 XP_011514762.1

View Full Product Details