Product Details

SNP ID
rs13396163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95035387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCAGCAGGTGTAGATGTGGTA[A/G]GCAGAATTTTCCATTCAAATGAAGA
Phenotype
MIM: 188860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAL PubMed Links

Gene Details

Gene
MAL
Gene Name
mal, T-cell differentiation protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002371.3 Intron NP_002362.1
NM_022438.2 Intron NP_071883.1
NM_022439.2 Intron NP_071884.1
NM_022440.2 Intron NP_071885.1

View Full Product Details