Product Details

SNP ID
rs9341314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:20579935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGAAGCATTTTTGTAAATATAC[G/T]TTCAGTATTTTGTGTGGAAGAACAC
Phenotype
MIM: 400014
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF1AY PubMed Links

Gene Details

Gene
EIF1AY
Gene Name
eukaryotic translation initiation factor 1A, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278612.1 Intron NP_001265541.1
NM_004681.3 Intron NP_004672.2

View Full Product Details