Product Details

SNP ID
rs13375579
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:153809781 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTTGAGTGGGGGAAGGTTTAACC[A/G]TCCACTGAACAAAGGGAGGACACAC
Phenotype
MIM: 614998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATAD2B PubMed Links

Gene Details

Gene
GATAD2B
Gene Name
GATA zinc finger domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005245364.4 2273 UTR 3 XP_005245421.1
Gene
LOC105371448
Gene Name
uncharacterized LOC105371448
There are no transcripts associated with this gene.

View Full Product Details