Product Details
- SNP ID
-
rs1332496
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
52
- Location
-
Chr.1:152709067 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGTCATTCAGGTTTATCGAAAT[C/T]CCACCAAGATGTCCTGCCAGCAGAA
- Phenotype
-
MIM: 612609
MIM: 612618
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LCE2A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147765240] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LCE2A
- Gene Name
- late cornified envelope 2A
There are no transcripts associated with this gene.
- Gene
- LCE4A
- Gene Name
- late cornified envelope 4A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178356.3 |
|
Intron |
|
|
NP_848133.1 |
View Full Product Details