Product Details
- SNP ID
-
rs11084213
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:52984559 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGACTGAATGGAGGAGGACGAACA[C/T]GGGAATGGAGTATGGAGTCAAAGAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF702P
PubMed Links
Gene Details
- Gene
- ZNF702P
- Gene Name
- zinc finger protein 702, pseudogene
There are no transcripts associated with this gene.
View Full Product Details