Product Details

SNP ID
rs12570038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80354669 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGATTAGATCATTAGTGTGGAGCC[G/T]TCATGCATGGGATTAATGCCCTTAT
Phenotype
MIM: 615154
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DYDC1 PubMed Links

Gene Details

Gene
DYDC1
Gene Name
DPY30 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001269053.1 Intron NP_001255982.1
NM_138812.3 Intron NP_620167.1
XM_005269549.4 Intron XP_005269606.1
XM_005269550.4 Intron XP_005269607.1
XM_011539334.2 Intron XP_011537636.1
XM_011539335.1 Intron XP_011537637.1
XM_017015749.1 Intron XP_016871238.1
Gene
DYDC2
Gene Name
DPY30 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270041.1 Intron NP_001256970.1
NM_001270042.1 Intron NP_001256971.1
NM_032372.5 Intron NP_115748.1
XM_011540267.2 Intron XP_011538569.1
XM_011540268.1 Intron XP_011538570.1
XM_011540270.2 Intron XP_011538572.1

View Full Product Details