Product Details
- SNP ID
-
rs12100749
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:55576217 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGATTTACACCTTGGGAGTCAA[C/T]ATGGTATTGGGGCACGTACATGGGA
- Phenotype
-
MIM: 600381
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KTN1
PubMed Links
Gene Details
- Gene
- KTN1
- Gene Name
- kinectin 1
There are no transcripts associated with this gene.
- Gene
- KTN1-AS1
- Gene Name
- KTN1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details