Product Details
- SNP ID
-
rs12948599
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:735633 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGAAAGCACCTGAGACCATAGA[G/T]GGGCATTGGAATAAGTATGGCATCT
- Phenotype
-
MIM: 611627
MIM: 606969
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM57A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115714847] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM57A
- Gene Name
- family with sequence similarity 57 member A
- Gene
- GEMIN4
- Gene Name
- gem nuclear organelle associated protein 4
There are no transcripts associated with this gene.
View Full Product Details