Product Details
- SNP ID
-
rs12066135
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:178849740 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATTTTGTTTAATTTGTTTCACCA[A/G]AAGTTTTATTTTGAGATGTGGCAGA
- Phenotype
-
MIM: 603874
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANGPTL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116610098] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANGPTL1
- Gene Name
- angiopoietin like 1
- Gene
- RALGPS2
- Gene Name
- Ral GEF with PH domain and SH3 binding motif 2
View Full Product Details