Product Details

SNP ID
rs12145411
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:179967094 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTCAGAGAGGCTTCTTAGACTA[C/T]ACTTTATACAATAGATCATCTCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP350 PubMed Links

Gene Details

Gene
CEP350
Gene Name
centrosomal protein 350
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014810.4 Intron NP_055625.4
XM_011510191.2 Intron XP_011508493.1
XM_011510198.2 Intron XP_011508500.1
XM_017002929.1 Intron XP_016858418.1
XM_017002930.1 Intron XP_016858419.1
XM_017002931.1 Intron XP_016858420.1

View Full Product Details