Product Details

SNP ID
rs12091190
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:197917878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTCGTGTCCTTTCCGCCCCCCA[A/G]CCATGCTCTTTCACGGGATCTCCGG
Phenotype
MIM: 606066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHX9 PubMed Links

Gene Details

Gene
LHX9
Gene Name
LIM homeobox 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014434.1 492 Intron NP_001014434.1
NM_020204.2 492 Missense Mutation ACC,GCC T19A NP_064589.2
XM_005245350.3 492 Intron XP_005245407.2
XM_011509781.2 492 Intron XP_011508083.2
XM_017001849.1 492 Intron XP_016857338.1

View Full Product Details