Product Details

SNP ID
rs10900542
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:204032569 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCCATGGCATCCACATTGTGACT[C/G]TAAGAAAAAGGCCAAGAAAAATACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details