Product Details

SNP ID
rs12077289
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:204037899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATAAAGAAATAGAAATGTGTGGG[G/T]TTTTTTCTTGTTTTTGATTGAAAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details