Product Details

SNP ID
rs13178795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:71719511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCGCCTTGTCTCTTCTCTTGCA[C/T]GCCTCCCTCCTCCCCCCACCCCCAC
Phenotype
MIM: 602606
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARTPT PubMed Links

Gene Details

Gene
CARTPT
Gene Name
CART prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004291.3 Intron NP_004282.1

View Full Product Details