Product Details

SNP ID
rs12821012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56222205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGCTTTGGAGGGGTGCAGGTGAC[A/T]TGTGTCTTAGAGCTGACGCAGATTG
Phenotype
MIM: 612104 MIM: 608730
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NABP2 PubMed Links

Gene Details

Gene
NABP2
Gene Name
nucleic acid binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024068.3 Intron NP_076973.1
XM_005269146.2 Intron XP_005269203.1
XM_005269147.2 Intron XP_005269204.1
XM_005269148.4 Intron XP_005269205.1
XM_005269149.4 Intron XP_005269206.1
Gene
RNF41
Gene Name
ring finger protein 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242826.1 Intron NP_001229755.1
NM_005785.3 Intron NP_005776.1
NM_194358.2 Intron NP_919339.1
NM_194359.2 Intron NP_919340.1
XM_005268561.4 Intron XP_005268618.1
XM_011537734.2 Intron XP_011536036.1
XM_011537735.2 Intron XP_011536037.1
Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
There are no transcripts associated with this gene.

View Full Product Details