Product Details

SNP ID
rs13401891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9955245 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTTTAATGAGTACTTTGACTTA[C/T]TGTGATTCGAAGCTAAAAATCTGTG
Phenotype
MIM: 609786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRHL1 PubMed Links

Gene Details

Gene
GRHL1
Gene Name
grainyhead like transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details