Product Details
- SNP ID
-
rs11960194
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:112985228 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTAATTACATATGTGTACATGCA[A/G]TATTAGTGTATATTAATATAAATGT
- Phenotype
-
MIM: 609844
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DCP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79979314] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DCP2
- Gene Name
- decapping mRNA 2
View Full Product Details