Product Details
- SNP ID
-
rs11701371
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42103862 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGATGTCTGCTGTTGCCTCTTCTT[C/G]GCTAGGTCGAGAGCTCCTTCCCACC
- Phenotype
-
MIM: 613859
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UMODL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116504428,rs146310440] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UMODL1
- Gene Name
- uromodulin like 1
- Gene
- UMODL1-AS1
- Gene Name
- UMODL1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details