Product Details

SNP ID
rs12031693
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:16199172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCGTCACTGGTCCAGGTCCTCAC[C/T]GACAGGATGTCAGTCTTCTCCAAGG
Phenotype
MIM: 612496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF19 PubMed Links

Gene Details

Gene
ARHGEF19
Gene Name
Rho guanine nucleotide exchange factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011540706.2 2530 Silent Mutation TCA,TCG S743S XP_011539008.1

View Full Product Details