Product Details
- SNP ID
-
rs12063995
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:151012404 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCCCTGTGTAATCATTAATAGCAC[C/T]CCTTTTCATTCTCATAAGCGTCCCA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM63A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80313919] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM63A
- Gene Name
- family with sequence similarity 63 member A
There are no transcripts associated with this gene.
- Gene
- PRUNE1
- Gene Name
- prune exopolyphosphatase
View Full Product Details