Product Details
- SNP ID
-
hCV31635540
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:17252176 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCATATTCTGTTCCTTCTTCTA[A/G]GTGTTGGAATCTTTGTGAAAATGTG
- Phenotype
-
MIM: 604665
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
COPS3
PubMed Links
Gene Details
- Gene
- COPS3
- Gene Name
- COP9 signalosome subunit 3
There are no transcripts associated with this gene.
View Full Product Details