Product Details

SNP ID
rs12732166
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:110660487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTTGAAGTGAATAAAAATCCA[A/C]CGCTAAGTGTCATGGAGAGAAAAAG
Phenotype
MIM: 176263
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNA3 PubMed Links

Gene Details

Gene
KCNA3
Gene Name
potassium voltage-gated channel subfamily A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002232.4 Intron NP_002223.3

View Full Product Details