Product Details

SNP ID
rs12485210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23391436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGTGCAGTGGCACAATCTCGGCT[C/T]ACGGCAAACTCTGCCTCCCAGATAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CES5AP1 PubMed Links

Gene Details

Gene
CES5AP1
Gene Name
carboxylesterase 5A pseudogene 1
There are no transcripts associated with this gene.

Gene
LOC107985555
Gene Name
coiled-coil domain-containing protein 188-like
There are no transcripts associated with this gene.

Gene
ZDHHC8P1
Gene Name
zinc finger DHHC-type containing 8 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details