Product Details

SNP ID
rs11196533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:113854915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGCCGCAGCCTCTCCGGCCTG[C/G]TCCCCGCGCAGACCTCGCTAGAGTA
Phenotype
MIM: 609682
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCLRE1A PubMed Links

Gene Details

Gene
DCLRE1A
Gene Name
DNA cross-link repair 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271816.1 284 Intron NP_001258745.1
NM_014881.4 284 Intron NP_055696.3
XM_006718090.1 284 Intron XP_006718153.1
XM_011540429.1 284 Intron XP_011538731.1
Gene
NHLRC2
Gene Name
NHL repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198514.3 284 Missense Mutation CTC,GTC L15V NP_940916.2
XM_011539769.2 284 Missense Mutation CTC,GTC L15V XP_011538071.1

View Full Product Details