Product Details

SNP ID
rs7916720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:90866810 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTACATCTTAAAGAGGTAGGAC[C/T]ACTCTTTATTTTCACAATCAAGCAT
Phenotype
MIM: 182137 MIM: 606115
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR7 PubMed Links

Gene Details

Gene
HTR7
Gene Name
5-hydroxytryptamine receptor 7
There are no transcripts associated with this gene.

Gene
RPP30
Gene Name
ribonuclease P/MRP subunit p30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104546.1 Intron NP_001098016.1
NM_006413.4 Intron NP_006404.1
XM_011539120.2 Intron XP_011537422.1
XM_011539121.2 Intron XP_011537423.1
XM_017015475.1 Intron XP_016870964.1
XM_017015476.1 Intron XP_016870965.1
XM_017015477.1 Intron XP_016870966.1
XM_017015478.1 Intron XP_016870967.1

View Full Product Details