Product Details

SNP ID
rs7638451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9946075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATCTCGAACTCCTGACCTCGTG[A/T]TCCACCCACCTCGGCCTCCCAAAGT
Phenotype
MIM: 607170
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CRELD1 PubMed Links

Gene Details

Gene
CRELD1
Gene Name
cysteine rich with EGF like domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031717.3 5114 Intron NP_001026887.1
NM_001077415.2 5114 Intron NP_001070883.1
NM_015513.4 5114 Intron NP_056328.2
XM_011534108.1 5114 Intron XP_011532410.1
XM_017007175.1 5114 Intron XP_016862664.1
Gene
PRRT3
Gene Name
proline rich transmembrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318871.1 5114 Intron NP_001305800.1
NM_207351.4 5114 UTR 3 NP_997234.3
XM_011533628.2 5114 UTR 3 XP_011531930.1
XM_011533629.2 5114 UTR 3 XP_011531931.1
XM_017006256.1 5114 UTR 3 XP_016861745.1
Gene
PRRT3-AS1
Gene Name
PRRT3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details