Product Details

SNP ID
rs7248424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1784087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGGCTCAGGCGTGTTTCATTGCAC[A/G]GATGGAGCGTGCCGTGTTGACCCAC
Phenotype
MIM: 605866 MIM: 611294
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8B3 PubMed Links

Gene Details

Gene
ATP8B3
Gene Name
ATPase phospholipid transporting 8B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178002.2 Intron NP_001171473.1
NM_138813.3 Intron NP_620168.1
XM_006722654.2 Intron XP_006722717.1
XM_006722655.2 Intron XP_006722718.1
XM_006722656.2 Intron XP_006722719.1
XM_011527707.1 Intron XP_011526009.1
XM_011527708.1 Intron XP_011526010.1
XM_011527709.1 Intron XP_011526011.1
XM_011527710.2 Intron XP_011526012.1
XM_011527711.2 Intron XP_011526013.1
XM_011527712.1 Intron XP_011526014.1
Gene
ONECUT3
Gene Name
one cut homeobox 3
There are no transcripts associated with this gene.

View Full Product Details