Product Details

SNP ID
rs11161572
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:85257146 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAATGGTCCAAAACATTAGTTCA[A/T]TTCAACAATTACTGAATACTTACAA
Phenotype
MIM: 603517
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BCL10 PubMed Links

Gene Details

Gene
BCL10
Gene Name
B-cell CLL/lymphoma 10
There are no transcripts associated with this gene.

Gene
C1orf52
Gene Name
chromosome 1 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198077.3 Intron NP_932343.1

View Full Product Details