Product Details

SNP ID
rs7933029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75773224 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAGCCTTCCTTGGGCCTCCTCTGT[A/C]CCTGTAACCACCTGTGCTAGGGACT
Phenotype
MIM: 606983
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DGAT2 PubMed Links

Gene Details

Gene
DGAT2
Gene Name
diacylglycerol O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253891.1 Intron NP_001240820.1
NM_032564.4 Intron NP_115953.2
XM_011545304.2 Intron XP_011543606.1
Gene
LOC283214
Gene Name
uncharacterized LOC283214
There are no transcripts associated with this gene.

View Full Product Details