Product Details

SNP ID
rs11696737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56393986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTCCTTCAGTGGTTAAGCTCA[G/T]GGATCTGGAGTTGGCAATTCTGGTG
Phenotype
MIM: 603072 MIM: 600369
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AURKA PubMed Links

Gene Details

Gene
AURKA
Gene Name
aurora kinase A
There are no transcripts associated with this gene.

Gene
CSTF1
Gene Name
cleavage stimulation factor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033521.1 Intron NP_001028693.1
NM_001033522.1 Intron NP_001028694.1
NM_001324.2 Intron NP_001315.1
XM_011528600.1 Intron XP_011526902.1

View Full Product Details