Product Details

SNP ID
rs12586352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:81186602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATAGAAGATGTAACTGAGATTT[C/T]TAATAAATGCAGAAAGAGAAGACAA
Phenotype
MIM: 600520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF2A1 PubMed Links

Gene Details

Gene
GTF2A1
Gene Name
general transcription factor IIA subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278940.1 Intron NP_001265869.1
NM_015859.3 Intron NP_056943.1
NM_201595.2 Intron NP_963889.1

View Full Product Details