Product Details
- SNP ID
-
rs12505890
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:1117867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGTAGCAAACGTAAAAGTTGCAT[C/T]TGACCTGTCCTCAGAAGCTATGCAA
- Phenotype
-
MIM: 612041
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105374344
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78628528] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105374344
- Gene Name
- uncharacterized LOC105374344
There are no transcripts associated with this gene.
- Gene
- RNF212
- Gene Name
- ring finger protein 212
There are no transcripts associated with this gene.
- Gene
- TMED11P
- Gene Name
- transmembrane p24 trafficking protein 11, pseudogene
There are no transcripts associated with this gene.
View Full Product Details