Product Details

SNP ID
rs6599293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:139246 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCAATGTTTTTTGTAGTAGTTTC[A/G]TAGTTTGATGTCTTAGATTTGTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF718 PubMed Links

Gene Details

Gene
ZNF718
Gene Name
zinc finger protein 718
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039127.5 Intron NP_001034216.2
NM_001289930.1 Intron NP_001276859.1
NM_001289931.1 Intron NP_001276860.1
XM_017007979.1 Intron XP_016863468.1
XM_017007980.1 Intron XP_016863469.1
XM_017007981.1 Intron XP_016863470.1

View Full Product Details