Product Details

SNP ID
rs13309357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128351949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCGGCAGGGGTGTGGCGCCC[A/T]TGAAGCCCGACCCTCCCAGGGGCGC
Phenotype
MIM: 612074
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRRT4 PubMed Links

Gene Details

Gene
PRRT4
Gene Name
proline rich transmembrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114726.2 2740 Intron NP_001108198.2
NM_001174164.1 2740 Missense Mutation AAG,ATG K536M NP_001167635.1
XM_005250342.3 2740 Missense Mutation AAG,ATG K536M XP_005250399.1
XM_005250343.3 2740 Missense Mutation AAG,ATG K536M XP_005250400.1
XM_006715985.3 2740 Missense Mutation AAG,ATG K536M XP_006716048.1
XM_017012205.1 2740 Missense Mutation AAG,ATG K536M XP_016867694.1
XM_017012206.1 2740 Intron XP_016867695.1
Gene
RBM28
Gene Name
RNA binding motif protein 28
There are no transcripts associated with this gene.

View Full Product Details