Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001006617.1 | 2780 | UTR 3 | NP_001006618.1 | ||
NM_001006618.1 | 2780 | Intron | NP_001006619.1 | ||
NM_001006619.1 | 2780 | UTR 3 | NP_001006620.1 | ||
NM_001006620.1 | 2780 | UTR 3 | NP_001006621.1 | ||
NM_001006621.1 | 2780 | UTR 3 | NP_001006622.1 | ||
NM_024117.3 | 2780 | UTR 3 | NP_077022.1 | ||
XM_011519005.1 | 2780 | UTR 3 | XP_011517307.1 | ||
XM_011519006.2 | 2780 | Intron | XP_011517308.1 | ||
XM_011519007.1 | 2780 | UTR 3 | XP_011517309.1 | ||
XM_011519008.1 | 2780 | Intron | XP_011517310.1 | ||
XM_011519009.1 | 2780 | Intron | XP_011517311.1 | ||
XM_011519010.2 | 2780 | Intron | XP_011517312.1 | ||
XM_017015126.1 | 2780 | UTR 3 | XP_016870615.1 | ||
XM_017015127.1 | 2780 | Intron | XP_016870616.1 |