Product Details

SNP ID
rs10849471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6500952 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAGCAGGGGAAGTGGTGATAAAA[C/T]GACTCAGGATATATTTTCAAAGTAG
Phenotype
MIM: 611855 MIM: 615638 MIM: 615639
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL51 PubMed Links

Gene Details

Gene
MRPL51
Gene Name
mitochondrial ribosomal protein L51
There are no transcripts associated with this gene.

Gene
NCAPD2
Gene Name
non-SMC condensin I complex subunit D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014865.3 Intron NP_055680.3
Gene
SCARNA10
Gene Name
small Cajal body-specific RNA 10
There are no transcripts associated with this gene.

View Full Product Details