Product Details
- SNP ID
-
rs11063285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:4653903 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGATTCTGTCCTTTCTTTTTTTTT[C/T]CCTCTTTTAAGACCTTTCTCAAAAC
- Phenotype
-
MIM: 604689
MIM: 603834
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKAP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113630331] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKAP3
- Gene Name
- A-kinase anchoring protein 3
There are no transcripts associated with this gene.
- Gene
- NDUFA9
- Gene Name
- NADH:ubiquinone oxidoreductase subunit A9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005002.4 |
|
Intron |
|
|
NP_004993.1 |
View Full Product Details