Product Details

SNP ID
rs11063251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4592528 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTGCAATCTGAGGTCCTTCCTG[A/G]TCCAGCTGCAGATTTGAGCGTGAAG
Phenotype
MIM: 609181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYRK4 PubMed Links

Gene Details

Gene
DYRK4
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282285.1 Intron NP_001269214.1
NM_001282286.1 Intron NP_001269215.1
NM_003845.2 Intron NP_003836.1

View Full Product Details