Product Details

SNP ID
rs7294327
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:63860856 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTTTAAAGAGAAAGGGTCT[C/T]GCTTTGTTGCCCAGACTGGAGTGCA
Phenotype
MIM: 606523
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SRGAP1 PubMed Links

Gene Details

Gene
SRGAP1
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020762.2 Intron NP_065813.1
XM_005269042.3 Intron XP_005269099.1
XM_011538580.1 Intron XP_011536882.1
XM_011538581.2 Intron XP_011536883.1

View Full Product Details