Product Details

SNP ID
hCV31846444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27644553 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAATTCACCTATTTTTTTTTA[A/T]AGATTTGTCTGGTTTTATTTTCTAA
Phenotype
MIM: 611479 MIM: 612762
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GPN1 PubMed Links

Gene Details

Gene
GPN1
Gene Name
GPN-loop GTPase 1
There are no transcripts associated with this gene.

Gene
SUPT7L
Gene Name
SPT7-like STAGA complex gamma subunit
There are no transcripts associated with this gene.

View Full Product Details