Product Details

SNP ID
rs11094147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70273953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAACAAACTAAAGCCTCCTGT[A/G]TGTCCTTCCCTGATCCAGTTTGCCT
Phenotype
MIM: 301770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARR3 PubMed Links

Gene Details

Gene
ARR3
Gene Name
arrestin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004312.2 Intron NP_004303.2
XM_017029518.1 Intron XP_016885007.1
Gene
RAB41
Gene Name
RAB41, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details