Product Details

SNP ID
rs12844904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70179429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGTGGCAGCAACCGTGTCAAA[A/T]CCGGGTCCCAGAAAGAGTGGGCAGA
Phenotype
MIM: 300926
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DGAT2L6 PubMed Links

Gene Details

Gene
DGAT2L6
Gene Name
diacylglycerol O-acyltransferase 2 like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198512.2 Intron NP_940914.1

View Full Product Details