Product Details

SNP ID
rs12029249
Assay Type
Functionally Tested
NCBI dbSNP Submissions
17
Location
Chr.1:35583539 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGTTCAGAAGTCAGACCATATCT[A/G]CCACCTGCGGACAGAGTGCCTTGTC
Phenotype
MIM: 614428
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TFAP2E PubMed Links

Gene Details

Gene
TFAP2E
Gene Name
transcription factor AP-2 epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178548.3 Intron NP_848643.2
XM_017001139.1 Intron XP_016856628.1

View Full Product Details