Product Details
- SNP ID
-
rs11580943
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:28236895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCTTCCTCGCCAGCACGCCTTA[A/G]CTTTGCAAGCCTGCATGCATTCAGG
- Phenotype
-
MIM: 614981
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATPIF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116668731] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATPIF1
- Gene Name
- ATPase inhibitory factor 1
- Gene
- DNAJC8
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C8
There are no transcripts associated with this gene.
- Gene
- LOC105376896
- Gene Name
- uncharacterized LOC105376896
There are no transcripts associated with this gene.
View Full Product Details